siRNA Details
Gene Symbol: Plekhg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081333.1 | NP_001074802.1 | 19 | 3395 | ||
GenBank mRNA | AK029859.1 | - | 17 | 4067 | ||
AK131125.1 | - | 13 | 4526 | |||
BC024792.1 | - | 3 | 848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
