siRNA Details
Gene Symbol: AAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014911.3 | NP_055726.3 | 2, 3 | 663 | ||
GenBank mRNA | AB028971.1 | - | 2, 3 | 542 | ||
AK056043.1 | - | 2, 3 | 470 | |||
AK310951.1 | - | 2, 3 | 466 | |||
BC002695.2 | - | 1, 2 | 198 | |||
BC104842.1 | - | 1, 2 | 182 | 8143845 | ||
BC111965.1 | - | 1, 2 | 182 | 8327433 | ||
BC143710.1 | - | 1, 2 | 182 | 9052224 | ||
DB285703.1 | - | 2, 3 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
