siRNA Details
Gene Symbol: Ibtk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081282.2 | NP_001074751.1 | 29 | 5284 | ||
GenBank mRNA | AK031796.1 | - | 18 | 3085 | ||
AK077387.1 | - | 7 | 1427 | |||
AK129353.1 | - | 26 | 4162 | |||
BC050255.1 | - | 17 | 4718 | |||
BC060722.1 | - | 28 | 5078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
