siRNA Details
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175375.3 | NP_780584.2 | 34 | 7906 | ||
GenBank mRNA | AK078916.1 | - | 6 | 1873 | ||
AK129284.1 | - | 2403 | ||||
BC091647.1 | - | 10 | 3331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
