siRNA Details
Gene Symbol: Trmt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089967.1 | - | 8 | 1412 | ||
BC053448.1 | - | 1 | 422 | |||
BC141035.1 | - | 8 | 1305 | 9055647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
