siRNA Details
Gene Symbol: Tmem132c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175432.3 | NP_780641.2 | 4 | 1390 | ||
GenBank mRNA | AK028289.1 | - | 4 | 1409 | ||
AK028541.1 | - | 4 | 1391 | |||
AK035053.1 | - | 2 | 288 | |||
AK157823.1 | - | 4 | 1391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
