siRNA Details
Gene Symbol: Mroh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162489.1 | NP_001155961.1 | 8 | 999 | ||
NM_175457.4 | NP_780666.3 | 9 | 1063 | |||
GenBank mRNA | AK036810.1 | - | 9 | 1064 | ||
AK038863.1 | - | 9 | 1045 | |||
AK052153.1 | - | 8 | 1015 | |||
AK090131.1 | - | 8 | 1399 | |||
AK122562.1 | - | 8 | 996 | |||
AK147808.1 | - | 9 | 1054 | |||
AK147897.1 | - | 9 | 1051 | |||
AK170702.1 | - | 9 | 1057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
