siRNA Details
Gene Symbol: Glis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080887.1 | - | 4, 5 | 583 | ||
AK087731.1 | - | 5, 6 | 799 | |||
AY220846.1 | - | 8, 9 | 2294 | |||
BC066143.1 | - | 9, 10 | 2629 | |||
BC129906.1 | - | 8, 9 | 2272 | |||
BC129907.1 | - | 8, 9 | 2272 | |||
DQ871027.1 | - | 10, 11 | 3249 |

More Information
Other Species Targeted

