siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 3 | 222 | ||
AK086119.1 | - | 2 | 195 | |||
AK147625.1 | - | 2 | 182 | |||
BB625805.1 | - | 3 | 215 | |||
BB662543.1 | - | 2 | 190 | |||
BC141190.1 | - | 2 | 268 | 9055802 | ||
BC171963.1 | - | 2 | 268 | 9054249 | ||
CJ109324.1 | - | 2 | 263 | |||
FJ628361.1 | - | 3 | 266 |
