siRNA Details
Gene Symbol: Huwe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021523.4 | NP_067498.4 | 7 | 980 | ||
GenBank mRNA | AK031374.1 | - | 8 | 1007 | ||
AK045183.1 | - | 7 | 918 | |||
AK049460.1 | - | 7 | 936 | |||
AK142956.1 | - | 8 | 966 | |||
AK163416.1 | - | 8 | 1130 | |||
AK165140.1 | - | 7 | 916 | |||
AK165648.1 | - | 8 | 1053 | |||
AY772010.3 | - | 7 | 917 | |||
AY929611.1 | - | 5 | 527 | |||
DQ097265.1 | - | 7 | 936 |

More Information
Other Species Targeted

Gene Ontology Categories:
