siRNA Details
Gene Symbol: Tanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181071.3 | NP_851416.2 | 21 | 3692 | ||
GenBank mRNA | AK028312.1 | - | 5 | 538 | ||
AK084601.1 | - | 4 | 467 | |||
AK173111.1 | - | 12 | 2162 | |||
BC089352.1 | - | 10 | 1683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
