siRNA Details
Gene Symbol: Apol7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024848.2 | NP_001020019.2 | 4, 5 | 381 | ||
GenBank mRNA | BC085284.1 | - | 4, 5 | 393 | 30750330 |
Gene Symbol: Apol7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164640.1 | NP_001158112.1 | 4, 5 | 487 | ||
NM_029419.2 | NP_083695.2 | 5, 6 | 587 | |||
GenBank mRNA | AK018646.1 | - | 5, 6 | 589 | ||
AK085956.1 | - | 4, 5 | 464 | |||
BC139137.1 | - | 4, 5 | 366 | 8862159 | ||
BC139138.1 | - | 4, 5 | 366 | 8862160 |
Gene Symbol: Apol7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175391.4 | NP_780600.1 | 4, 5 | 405 | ||
GenBank mRNA | AK075855.1 | - | 4, 5 | 407 | ||
BC139148.1 | - | 4, 5 | 364 | 8862170 | ||
BC139149.1 | - | 4, 5 | 364 | 8862171 |
Gene Symbol: Apol7e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134802.1 | NP_001128274.1 | 4, 5 | 381 | ||
GenBank mRNA | AK134162.1 | - | 4, 5 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
