siRNA Details
Gene Symbol: Sobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175407.3 | NP_780616.4 | 7 | 3453 | ||
GenBank mRNA | AK077362.1 | - | 2 | 480 | ||
BC058972.1 | - | 2 | 2315 | |||
BC059851.1 | - | 7 | 3453 | 5704623 | ||
DQ157775.1 | - | 7 | 3012 |

More Information
Other Species Targeted

Gene Ontology Categories:
