siRNA Details
Gene Symbol: Zfp526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175436.4 | NP_780645.1 | 3 | 886 | ||
GenBank mRNA | AK050839.1 | - | 3 | 887 | ||
AK051798.1 | - | 3 | 883 | |||
AK173308.1 | - | 796 | ||||
BC052548.1 | - | 2 | 840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
