siRNA Details
Gene Symbol: Rbbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081223.2 | NP_001074692.1 | 3, 4 | 591 | ||
NM_001252495.1 | NP_001239424.1 | 3, 4 | 496 | |||
GenBank mRNA | AK037049.1 | - | 3, 4 | 513 | ||
AK048254.1 | - | 3, 4 | 533 | |||
AK077614.1 | - | 3, 4 | 532 | |||
AK163908.1 | - | 558 | ||||
BC027367.1 | - | 3, 4 | 459 | |||
BC050849.1 | - | 3, 4 | 496 | 6508067 | ||
CD352622.1 | - | 2, 3 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
