siRNA Details
Gene Symbol: Glis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080887.1 | - | 5 | 859 | ||
AK087731.1 | - | 6 | 1075 | |||
BC066143.1 | - | 10 | 2905 | |||
DQ871027.1 | - | 11 | 3525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
