siRNA Details
Gene Symbol: Kcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033953.1 | - | 4 | 302 | ||
AK086119.1 | - | 3 | 275 | |||
AK147625.1 | - | 3 | 262 | |||
BB625805.1 | - | 4 | 295 | |||
BB662543.1 | - | 3 | 270 | |||
BC141190.1 | - | 3 | 348 | 9055802 | ||
BC171963.1 | - | 3 | 348 | 9054249 | ||
BY261633.1 | - | 3 | 363 | |||
CJ109324.1 | - | 3 | 343 | |||
FJ628361.1 | - | 4 | 346 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_021853, XM_006233706, XM_006233699, XM_006233702, XM_006233700, and others.
