siRNA Details
Gene Symbol: Lrfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027452.3 | NP_081728.2 | 2 | 1112 | ||
GenBank mRNA | AK017594.1 | - | 2 | 1316 | ||
AK044375.1 | - | 2 | 989 | |||
AK122476.1 | - | 2 | 1067 | |||
BC139418.1 | - | 2 | 1112 | 8862440 |
Gene Symbol: Lrfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175478.2 | NP_780687.1 | 2 | 1728 | ||
GenBank mRNA | AK040916.1 | - | 2 | 1729 | ||
BC066999.1 | - | 3 | 1651 | 6418622 | ||
BC094593.1 | - | 2 | 1690 | 5708704 | ||
DQ078787.1 | - | 1 | 872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
