siRNA Details
Gene Symbol: Huwe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021523.4 | NP_067498.4 | 8 | 1088 | ||
GenBank mRNA | AK031374.1 | - | 9 | 1115 | ||
AK037281.1 | - | 1 | 1303 | |||
AK045183.1 | - | 8 | 1026 | |||
AK049460.1 | - | 8 | 1044 | |||
AK142956.1 | - | 9 | 1075 | |||
AK163416.1 | - | 9 | 1238 | |||
AK165140.1 | - | 8 | 1024 | |||
AK165648.1 | - | 9 | 1161 | |||
AY772010.3 | - | 8 | 1025 | |||
AY929611.1 | - | 6 | 635 | |||
DQ097265.1 | - | 8 | 1044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
