siRNA Details
Gene Symbol: Tanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181071.3 | NP_851416.2 | 21 | 3724 | ||
GenBank mRNA | AK028312.1 | - | 5 | 570 | ||
AK084601.1 | - | 4 | 499 | |||
AK173111.1 | - | 12 | 2194 | |||
BC089352.1 | - | 10 | 1715 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191653, XM_008768355, XM_008768352, XM_008768351, XM_008768354, and others.
Gene Ontology Categories:
