siRNA Details
Gene Symbol: Fam73b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242407.1 | NP_001229336.1 | 16 | 2053 | ||
NM_175392.3 | NP_780601.1 | 16 | 2221 | |||
GenBank mRNA | AK077618.1 | - | 16 | 2014 | ||
AK131152.1 | - | 12 | 3795 | |||
AK154200.1 | - | 16 | 2042 | |||
AK165996.1 | - | 16 | 2045 | |||
BC038823.1 | - | 4 | 463 | |||
BC051404.1 | - | 16 | 2221 | 6395030 |
