siRNA Details
Gene Symbol: CHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005273.2 | NP_001005273.1 | 1, 2 | 250 | ||
NM_005852.3 | NP_005843.2 | 1, 2 | 250 | |||
XM_017024063.1 | XP_016879552.1 | 1, 2 | 235 | |||
XM_017024066.1 | XP_016879555.1 | 2, 3 | 55 | |||
XM_017024067.1 | XP_016879556.1 | 2, 3 | 238 | |||
GenBank mRNA | U91543.1 | - | 1, 2 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
