siRNA Details
Gene Symbol: Zfp612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175480.4 | NP_780689.3 | 5 | 2949 | ||
GenBank mRNA | AK043395.1 | - | 1 | 1134 | ||
AK162867.1 | - | 1 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
