siRNA Details
Gene Symbol: AAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028971.1 | - | 18 | 2975 | ||
BC104842.1 | - | 17 | 2615 | 8143845 | ||
BC111965.1 | - | 17 | 2615 | 8327433 | ||
BC143710.1 | - | 17 | 2615 | 9052224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
