siRNA Details
Gene Symbol: Adamts6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030315.1 | - | 20 | 3882 | ||
AK041499.1 | - | 1 | 160 | |||
AK044528.1 | - | 20 | 3712 | |||
BC172116.1 | - | 22 | 3038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
