siRNA Details
Gene Symbol: Pgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163314.2 | NP_001156786.1 | 1 | 211 | ||
GenBank mRNA | AK041133.1 | - | 1 | 212 | ||
AK051360.1 | - | 1 | 155 | |||
AK051866.1 | - | 1 | 222 | |||
AK138171.1 | - | 1 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
