siRNA Details
Gene Symbol: C130060K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175524.4 | NP_780733.2 | 2 | 440 | ||
GenBank mRNA | AK048439.1 | - | 2 | 350 | ||
AK051723.1 | - | 2 | 374 | |||
BC119578.1 | - | 2 | 341 | 40058179 | ||
BC119579.1 | - | 2 | 341 | 40058180 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
