siRNA Details
Gene Symbol: Scaper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081341.1 | NP_001074810.1 | 5 | 346 | ||
GenBank mRNA | AK031358.1 | - | 5 | 332 | ||
AK048755.1 | - | 5 | 353 | |||
AK049219.1 | - | 4 | 310 | |||
AK049497.1 | - | 5 | 342 | |||
AK173175.1 | - | 4 | 294 | |||
BC095992.1 | - | 4 | 294 | |||
BC157959.1 | - | 4 | 309 | 9007286 | ||
BC158062.1 | - | 4 | 286 | 9088165 | ||
BC172119.1 | - | 4 | 309 | 9054405 | ||
BC172151.1 | - | 4 | 286 | 9054821 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
