siRNA Details
Gene Symbol: Nlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081280.1 | NP_001074749.1 | 7 | 2579 | ||
NM_175547.3 | NP_780756.1 | 4 | 502 | |||
GenBank mRNA | AK079766.1 | - | 4 | 426 | ||
AK084199.1 | - | 4 | 416 | |||
AK171971.1 | - | 6 | 3592 | |||
BC130223.1 | - | 4 | 1952 | |||
BC152543.1 | - | 4 | 1952 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
