siRNA Details
Gene Symbol: Tmem169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175564.4 | NP_780773.1 | 2 | 359 | ||
GenBank mRNA | AK034935.1 | - | 2 | 364 | ||
AK043681.1 | - | 2 | 360 | |||
AK084021.1 | - | 2 | 338 | |||
AK149157.1 | - | 1 | 669 | |||
BC116730.1 | - | 1 | 257 | 40126049 | ||
BC117087.1 | - | 1 | 257 | 40126406 |

More Information
Other Species Targeted

Gene Ontology Categories:
