siRNA Details
Gene Symbol: Olfr212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011800.2 | NP_001011800.1 | 2 | 777 | ||
GenBank mRNA | AK029829.1 | - | 2 | 779 | ||
BC141000.1 | - | 2 | 586 | 9055612 |
Gene Symbol: Olfr213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011801.1 | NP_001011801.1 | 1 | 292 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
