siRNA Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031075.1 | - | 3 | 3354 | ||
AK038524.1 | - | 1 | 236 | |||
AK038736.1 | - | 1 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
