siRNA Details
Gene Symbol: Adamts6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081020.1 | NP_001074489.1 | 22 | 3200 | ||
GenBank mRNA | AK030315.1 | - | 22 | 4138 | ||
AK034499.1 | - | 5 | 585 | |||
AK041499.1 | - | 3 | 416 | |||
BC172116.1 | - | 24 | 3294 |

More Information
Other Species Targeted

Gene Ontology Categories:
