siRNA Details
Gene Symbol: APITD1-CORT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243768.1 | NP_001230697.1 | 4 | 783 | ||
NM_001270517.1 | NP_001257446.1 | 6 | 896 | |||
NM_198544.3 | NP_940946.1 | 5 | 719 | |||
NM_199006.2 | NP_950171.2 | 4 | 652 | |||
NR_037187.1 | 3 | 749 | ||||
GenBank mRNA | AF516753.1 | - | 4 | 370 | ||
AF521016.1 | - | 5 | 719 | |||
CF128105.1 | - | 4 | 722 |
Gene Symbol: CORT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302.4 | NP_001293.3 | 2 | 631 | ||
GenBank mRNA | AB000263.1 | - | 2 | 131 | ||
AF013252.1 | - | 2 | 211 | |||
AF086433.1 | - | 2 | 436 | |||
AY358561.1 | - | 2 | 631 | |||
BC040034.1 | - | 2 | 179 | |||
BC119724.1 | - | 2 | 311 | 40118115 | ||
BC119725.1 | - | 2 | 311 | 40118117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
