siRNA Details
Gene Symbol: Nlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081280.1 | NP_001074749.1 | 8 | 2707 | ||
NM_175547.3 | NP_780756.1 | 5 | 630 | |||
GenBank mRNA | AK079766.1 | - | 5 | 554 | ||
AK084199.1 | - | 5 | 544 | |||
AK171971.1 | - | 7 | 3720 | |||
BC130223.1 | - | 5 | 2080 | |||
BC152543.1 | - | 5 | 2080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
