siRNA Details
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175550.3 | NP_780759.2 | 2 | 240 | ||
GenBank mRNA | AK036944.1 | - | 2 | 223 | ||
AK039257.1 | - | 2 | 228 | |||
AK143351.1 | - | 2 | 230 | |||
AK144636.1 | - | 2 | 220 | |||
AK163701.1 | - | 2 | 231 | |||
BY037321.1 | - | 2 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
