siRNA Details
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166584.1 | NP_001160056.1 | 14 | 8001 | ||
NM_001166585.1 | NP_001160057.1 | 11 | 7519 | |||
NM_009346.3 | NP_033372.1 | 13 | 7938 | |||
GenBank mRNA | AK045406.1 | - | 1 | 1885 | ||
BC027158.1 | - | 1 | 746 | |||
BC049153.1 | - | 1 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
