siRNA Details
Gene Symbol: Trim30b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175648.2 | NP_783579.1 | 2 | 1448 | ||
GenBank mRNA | AK040770.1 | - | 2176 | |||
AK041488.1 | - | 2 | 1448 | |||
AK080092.1 | - | 2177 | ||||
AK162946.1 | - | 3882 |
