siRNA Details
Gene Symbol: Cnpy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175651.4 | NP_783582.1 | 3 | 422 | ||
GenBank mRNA | AK035828.1 | - | 3 | 423 | ||
AK035938.1 | - | 2 | 962 | |||
AK139155.1 | - | 2 | 1108 | |||
AK162570.1 | - | 2 | 911 | |||
BC096464.1 | - | 347 | 6390940 |
