siRNA Details
Gene Symbol: Zfp738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001187.3 | NP_001001187.2 | 4 | 2357 | ||
GenBank mRNA | AK032995.1 | - | 1 | 897 | ||
BC082605.1 | - | 4 | 2312 |
Gene Symbol: Zfp729a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183146.3 | NP_898969.2 | 4 | 1935 | ||
GenBank mRNA | AK040978.1 | - | 4 | 1976 | ||
AK138962.1 | - | 4 | 1935 | |||
BC098180.1 | - | 4 | 1947 | 5401052 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
