siRNA Details
Gene Symbol: Adamts6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081020.1 | NP_001074489.1 | 23 | 3349 | ||
GenBank mRNA | AK030315.1 | - | 23 | 4287 | ||
AK034499.1 | - | 6 | 734 | |||
AK041499.1 | - | 4 | 565 | |||
BC172116.1 | - | 25 | 3443 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008760707.
Gene Ontology Categories:
