siRNA Details
Gene Symbol: E330021D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201390.1 | NP_001188319.1 | 2 | 832 | ||
GenBank mRNA | AK054382.1 | - | 2 | 833 | ||
AK135843.1 | - | 2 | 833 | |||
AK162229.1 | - | 2 | 762 | |||
BC099944.1 | - | 2 | 823 |
