siRNA Details
Gene Symbol: CSTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033505.1 | NP_001028677.1 | 2, 3 | 297 | ||
NM_001326.2 | NP_001317.1 | 2, 3 | 297 | |||
GenBank mRNA | AK290836.1 | - | 2, 3 | 297 | ||
BC009792.1 | - | 2, 3 | 279 | |||
BC059948.1 | - | 2, 3 | 151 | |||
BC108319.1 | - | 2, 3 | 271 | 6197136 | ||
U15782.1 | - | 2, 3 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
