siRNA Details
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175550.3 | NP_780759.2 | 3 | 282 | ||
GenBank mRNA | AK036944.1 | - | 3 | 265 | ||
AK039257.1 | - | 3 | 270 | |||
AK143351.1 | - | 3 | 272 | |||
AK144636.1 | - | 3 | 262 | |||
BY037321.1 | - | 3 | 283 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191742.
Gene Ontology Categories:
