siRNA Details
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113209.2 | NP_001106680.1 | 12 | 7963 | ||
NM_001113210.2 | NP_001106681.1 | 12 | 7936 | |||
NM_001286127.1 | NP_001273056.1 | 11 | 7847 | |||
NM_001286131.1 | NP_001273060.1 | 10 | 7714 | |||
NM_008687.6 | NP_032713.3 | 9 | 7625 | |||
GenBank mRNA | AK030686.1 | - | 1 | 1171 | ||
AK050276.1 | - | 1 | 2208 | |||
AK053181.1 | - | 1 | 4012 | |||
AK081555.1 | - | 1 | 4013 | |||
BC031188.1 | - | 1 | 973 | |||
BC096542.1 | - | 1 | 1330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
