siRNA Details
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166584.1 | NP_001160056.1 | 14 | 8091 | ||
NM_001166585.1 | NP_001160057.1 | 11 | 7609 | |||
NM_009346.3 | NP_033372.1 | 13 | 8028 | |||
GenBank mRNA | AK035191.1 | - | 1 | 1655 | ||
AK045406.1 | - | 1 | 1975 | |||
BC027158.1 | - | 1 | 836 | |||
BC049153.1 | - | 1 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
