siRNA Details
Gene Symbol: Ikzf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011772.2 | NP_035902.2 | 8 | 3220 | ||
GenBank mRNA | AK041640.1 | - | 1 | 950 | ||
AK089522.1 | - | 9 | 1854 | |||
AK170453.1 | - | 9 | 3144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
