siRNA Details
Gene Symbol: Zfp608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175751.4 | NP_786927.2 | 5 | 3483 | ||
GenBank mRNA | AK076178.1 | - | 1 | 359 | ||
AK077030.1 | - | 4 | 2210 | |||
BC059268.1 | - | 4 | 3150 | |||
BC092219.1 | - | 5 | 3216 | 6830664 | ||
BC100388.1 | - | 1 | 732 | |||
BC151073.1 | - | 4 | 3002 | 9087986 | ||
BC151075.1 | - | 4 | 3002 | 9087988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
