siRNA Details
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146003.2 | NP_666115.2 | 2 | 703 | ||
GenBank mRNA | AK053904.1 | - | 2 | 703 | ||
AK083377.1 | - | 2 | 292 | |||
BC052718.1 | - | 2 | 507 | |||
BC061480.1 | - | 2 | 527 | 6509771 | ||
BC151128.1 | - | 2 | 113 | 9088041 | ||
BC172171.1 | - | 2 | 113 | 9054841 |

More Information
Other Species Targeted

Gene Ontology Categories:
