siRNA Details
Gene Symbol: A330041J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039418.1 | - | 2, 3 | 318 | ||
AK046134.1 | - | 2, 3 | 234 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_176941.1 | - | 318 | Chr. - - [-] on Build |
